siRNA Details
| Gene Symbol: Olfr1121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146348.2 | NP_666460.2 | 1 | 601 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1121| Gene Symbol: Olfr1121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146348.2 | NP_666460.2 | 1 | 601 | ||