siRNA Details
| Gene Symbol: Olfr1128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146349.2 | NP_666461.2 | 1 | 412 | ||
| GenBank mRNA | BC104064.2 | - | 1 | 411 | ||
| BC104065.1 | - | 1 | 412 | 40041018 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1128| Gene Symbol: Olfr1128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146349.2 | NP_666461.2 | 1 | 412 | ||
| GenBank mRNA | BC104064.2 | - | 1 | 411 | ||
| BC104065.1 | - | 1 | 412 | 40041018 | ||