siRNA Details
Gene Symbol: Olfr1134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147030.2 | NP_667241.2 | 2 | 408 | ||
GenBank mRNA | BC125403.1 | - | 1 | 301 | 40129918 | |
BC125405.1 | - | 1 | 301 | 40129920 |
Gene Symbol: Olfr1133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146351.2 | NP_666463.2 | 1 | 281 | ||
GenBank mRNA | BC104086.1 | - | 1 | 281 | ||
BC104087.2 | - | 1 | 280 |
