siRNA Details
| Gene Symbol: Olfr692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146355.1 | NP_666467.1 | 1 | 937 | ||
| GenBank mRNA | BC119096.1 | - | 1 | 1035 | 8733845 | |
| BC120788.1 | - | 1 | 1035 | 40129711 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr692| Gene Symbol: Olfr692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146355.1 | NP_666467.1 | 1 | 937 | ||
| GenBank mRNA | BC119096.1 | - | 1 | 1035 | 8733845 | |
| BC120788.1 | - | 1 | 1035 | 40129711 | ||