siRNA Details
| Gene Symbol: Olfr168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146357.1 | NP_666469.1 | 1 | 722 | ||
| GenBank mRNA | BC127969.1 | - | 1 | 726 | 40135070 | |
| Gene Symbol: Olfr166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147068.1 | NP_667279.1 | 1 | 722 | ||
| GenBank mRNA | BC116856.1 | - | 1 | 769 | 40126175 | |
| BC119189.1 | - | 1 | 769 | 8733938 | ||

Olfr168
Genomic Map