siRNA Details
Gene Symbol: Olfr574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146360.2 | NP_666472.2 | 1 | 868 | ||
GenBank mRNA | BC127050.1 | - | 1 | 866 | 40081239 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146360.2 | NP_666472.2 | 1 | 868 | ||
GenBank mRNA | BC127050.1 | - | 1 | 866 | 40081239 |