siRNA Details
| Gene Symbol: Olfr574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146360.2 | NP_666472.2 | 1 | 480 | ||
| GenBank mRNA | BC127050.1 | - | 1 | 478 | 40081239 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr574| Gene Symbol: Olfr574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146360.2 | NP_666472.2 | 1 | 480 | ||
| GenBank mRNA | BC127050.1 | - | 1 | 478 | 40081239 | |