siRNA Details
Gene Symbol: Olfr557 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146361.2 | NP_666473.2 | 1 | 834 | ||
GenBank mRNA | BC145899.1 | - | 1 | 834 | 40131202 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr557 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146361.2 | NP_666473.2 | 1 | 834 | ||
GenBank mRNA | BC145899.1 | - | 1 | 834 | 40131202 |