siRNA Details
| Gene Symbol: Olfr1094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146365.2 | NP_666477.1 | 1 | 554 | ||
| GenBank mRNA | BC120867.1 | - | 1 | 554 | 40129790 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1094| Gene Symbol: Olfr1094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146365.2 | NP_666477.1 | 1 | 554 | ||
| GenBank mRNA | BC120867.1 | - | 1 | 554 | 40129790 | |
Set Membership: |
|