siRNA Details
| Gene Symbol: Olfr434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146369.1 | NP_666481.1 | 1 | 70 | ||
| GenBank mRNA | BC120611.1 | - | 1 | 102 | 40129534 | |
| BC120613.1 | - | 1 | 102 | 40129536 | ||
| Gene Symbol: Olfr47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146370.1 | NP_666482.1 | 1 | 70 | ||
| GenBank mRNA | BC120658.1 | - | 1 | 102 | 40129581 | |
| BC120660.1 | - | 1 | 102 | 40129583 | ||

Olfr434
Genomic Map