siRNA Details
| Gene Symbol: Olfr918 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146375.2 | NP_666487.2 | 1 | 432 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr918| Gene Symbol: Olfr918 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146375.2 | NP_666487.2 | 1 | 432 | ||