siRNA Details
| Gene Symbol: Olfr147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146869.2 | NP_667080.1 | 2 | 837 | ||
| Gene Symbol: Olfr918 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146375.2 | NP_666487.2 | 1 | 646 | ||

Olfr147
Genomic Map