siRNA Details
| Gene Symbol: Olfr323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146376.2 | NP_666488.2 | 1 | 913 | ||
| GenBank mRNA | BC146611.1 | - | 2 | 353 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr323| Gene Symbol: Olfr323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146376.2 | NP_666488.2 | 1 | 913 | ||
| GenBank mRNA | BC146611.1 | - | 2 | 353 | ||