siRNA Details
| Gene Symbol: Olfr127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146377.1 | NP_666489.1 | 1 | 914 | ||
| GenBank mRNA | BC132433.1 | - | 1 | 940 | 40130710 | |
| BC145997.1 | - | 1 | 940 | 40131300 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr127| Gene Symbol: Olfr127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146377.1 | NP_666489.1 | 1 | 914 | ||
| GenBank mRNA | BC132433.1 | - | 1 | 940 | 40130710 | |
| BC145997.1 | - | 1 | 940 | 40131300 | ||