siRNA Details
Gene Symbol: Olfr1350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146389.1 | NP_666501.1 | 1 | 125 | ||
GenBank mRNA | BC104062.1 | - | 1 | 124 | ||
BC104063.1 | - | 1 | 125 | 40040754 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146389.1 | NP_666501.1 | 1 | 125 | ||
GenBank mRNA | BC104062.1 | - | 1 | 124 | ||
BC104063.1 | - | 1 | 125 | 40040754 |