siRNA Details
| Gene Symbol: Olfr1277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146396.1 | NP_666508.1 | 1 | 651 | ||
| GenBank mRNA | BC120803.1 | - | 1 | 678 | 40129726 | |
| BC120805.1 | - | 1 | 674 | 40129728 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1277| Gene Symbol: Olfr1277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146396.1 | NP_666508.1 | 1 | 651 | ||
| GenBank mRNA | BC120803.1 | - | 1 | 678 | 40129726 | |
| BC120805.1 | - | 1 | 674 | 40129728 | ||