siRNA Details
| Gene Symbol: Olfr1328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146399.2 | NP_666511.2 | 1 | 681 | ||
| GenBank mRNA | BC106978.2 | - | 1 | 681 | 40042180 | |
| BC148290.2 | - | 1 | 681 | 40042169 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1328| Gene Symbol: Olfr1328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146399.2 | NP_666511.2 | 1 | 681 | ||
| GenBank mRNA | BC106978.2 | - | 1 | 681 | 40042180 | |
| BC148290.2 | - | 1 | 681 | 40042169 | ||