siRNA Details
| Gene Symbol: Olfr1289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146404.1 | NP_666516.1 | 1 | 327 | ||
| GenBank mRNA | BC120736.1 | - | 1 | 409 | 40129659 | |
| BC137761.1 | - | 1 | 410 | 8860780 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1289| Gene Symbol: Olfr1289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146404.1 | NP_666516.1 | 1 | 327 | ||
| GenBank mRNA | BC120736.1 | - | 1 | 409 | 40129659 | |
| BC137761.1 | - | 1 | 410 | 8860780 | ||