siRNA Details
| Gene Symbol: Olfr228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146405.2 | NP_666517.2 | 1 | 524 | ||
| Gene Symbol: Olfr1076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146406.2 | NP_666518.2 | 1 | 524 | ||

Olfr228
Genomic Map