siRNA Details
| Gene Symbol: Olfr1080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146409.2 | NP_666521.2 | 2 | 447 | ||
| GenBank mRNA | CB174209.1 | - | 2 | 447 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1080| Gene Symbol: Olfr1080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146409.2 | NP_666521.2 | 2 | 447 | ||
| GenBank mRNA | CB174209.1 | - | 2 | 447 | ||