siRNA Details
| Gene Symbol: Olfr1420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146410.1 | NP_666522.1 | 1 | 376 | ||
| GenBank mRNA | BC120832.1 | - | 1 | 400 | 40129755 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1420| Gene Symbol: Olfr1420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146410.1 | NP_666522.1 | 1 | 376 | ||
| GenBank mRNA | BC120832.1 | - | 1 | 400 | 40129755 | |