siRNA Details
| Gene Symbol: Olfr464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146412.2 | NP_666524.2 | 1 | 228 | ||
| GenBank mRNA | BC120701.1 | - | 1 | 229 | 40129624 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr464| Gene Symbol: Olfr464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146412.2 | NP_666524.2 | 1 | 228 | ||
| GenBank mRNA | BC120701.1 | - | 1 | 229 | 40129624 | |