siRNA Details
| Gene Symbol: Olfr881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146418.2 | NP_666530.1 | 1 | 830 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr881| Gene Symbol: Olfr881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146418.2 | NP_666530.1 | 1 | 830 | ||