siRNA Details
| Gene Symbol: Olfr888 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146424.1 | NP_666635.1 | 1 | 827 | ||
| Gene Symbol: Olfr890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146481.2 | NP_666692.2 | 1 | 827 | ||

Olfr888
Genomic Map