siRNA Details
| Gene Symbol: THAP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004705.3 | NP_004696.2 | 2, 3 | 338 | ||
| GenBank mRNA | AF007393.1 | - | 2, 3 | 209 | ||
| AF081567.1 | - | 2, 3 | 209 | |||
| AK291843.1 | - | 2, 3 | 210 | |||
| BC110304.1 | - | 2, 3 | 315 | 5222582 | ||
| BC117138.1 | - | 2, 3 | 267 | 40125689 | ||
| BC117140.1 | - | 2, 3 | 267 | 40125691 | ||

THAP12
Genomic Map