siRNA Details
Gene Symbol: Olfr223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146429.1 | NP_666640.1 | 1 | 778 | ||
GenBank mRNA | BC119106.1 | - | 1 | 868 | 8733855 | |
BC119108.1 | - | 1 | 868 | 8733857 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146429.1 | NP_666640.1 | 1 | 778 | ||
GenBank mRNA | BC119106.1 | - | 1 | 868 | 8733855 | |
BC119108.1 | - | 1 | 868 | 8733857 |