siRNA Details
| Gene Symbol: THAP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004705.3 | NP_004696.2 | 3 | 369 | ||
| NR_130898.1 | 4 | 464 | ||||
| GenBank mRNA | AF007393.1 | - | 3 | 240 | ||
| AF081567.1 | - | 3 | 240 | |||
| AK291843.1 | - | 3 | 241 | |||
| BC110304.1 | - | 3 | 346 | 5222582 | ||
| BC117138.1 | - | 3 | 298 | 40125689 | ||
| BC117140.1 | - | 3 | 298 | 40125691 | ||
| DR005363.1 | - | 4 | 470 | |||

THAP12
Genomic Map