siRNA Details
| Gene Symbol: Olfr998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146436.2 | NP_666647.2 | 1 | 826 | ||
| GenBank mRNA | BC150719.1 | - | 1 | 826 | 9087630 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr998| Gene Symbol: Olfr998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146436.2 | NP_666647.2 | 1 | 826 | ||
| GenBank mRNA | BC150719.1 | - | 1 | 826 | 9087630 | |