siRNA Details
Gene Symbol: Olfr998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146436.2 | NP_666647.2 | 1 | 95 | ||
GenBank mRNA | BC150719.1 | - | 1 | 95 | 9087630 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146436.2 | NP_666647.2 | 1 | 95 | ||
GenBank mRNA | BC150719.1 | - | 1 | 95 | 9087630 |