siRNA Details
Gene Symbol: PRKX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005044.4 | NP_005035.1 | 6 | 1237 | ||
GenBank mRNA | BC041073.1 | - | 6 | 1040 | 5485308 | |
X85545.1 | - | 6 | 1222 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PRKX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005044.4 | NP_005035.1 | 6 | 1237 | ||
GenBank mRNA | BC041073.1 | - | 6 | 1040 | 5485308 | |
X85545.1 | - | 6 | 1222 |