siRNA Details
| Gene Symbol: Olfr938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146438.1 | NP_666649.1 | 1 | 517 | ||
| GenBank mRNA | BC125399.1 | - | 1 | 590 | 40129914 | |
| BC125401.1 | - | 1 | 590 | 40129916 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr938| Gene Symbol: Olfr938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146438.1 | NP_666649.1 | 1 | 517 | ||
| GenBank mRNA | BC125399.1 | - | 1 | 590 | 40129914 | |
| BC125401.1 | - | 1 | 590 | 40129916 | ||