siRNA Details
Gene Symbol: Olfr919 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146440.1 | NP_666651.1 | 1 | 905 |
Gene Symbol: Olfr150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146609.2 | NP_666820.2 | 1 | 893 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
