siRNA Details
| Gene Symbol: Olfr933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146441.1 | NP_666652.1 | 1 | 867 | ||
| GenBank mRNA | BC119155.1 | - | 1 | 894 | 8733904 | |
| BC119157.1 | - | 1 | 894 | 8733906 | ||
| Gene Symbol: Olfr930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146272.1 | NP_666384.1 | 1 | 867 | ||
| GenBank mRNA | BC119252.1 | - | 1 | 949 | 8734001 | |
| BC119254.1 | - | 1 | 951 | 8734003 | ||

Olfr933
Genomic Map