siRNA Details
| Gene Symbol: Olfr933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146441.1 | NP_666652.1 | 1 | 594 | ||
| GenBank mRNA | BC119155.1 | - | 1 | 621 | 8733904 | |
| BC119157.1 | - | 1 | 621 | 8733906 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr933| Gene Symbol: Olfr933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146441.1 | NP_666652.1 | 1 | 594 | ||
| GenBank mRNA | BC119155.1 | - | 1 | 621 | 8733904 | |
| BC119157.1 | - | 1 | 621 | 8733906 | ||