siRNA Details
| Gene Symbol: Olfr933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146441.1 | NP_666652.1 | 1 | 391 | ||
| GenBank mRNA | BC119155.1 | - | 1 | 418 | 8733904 | |
| BC119157.1 | - | 1 | 418 | 8733906 | ||
| Gene Symbol: Olfr26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146783.2 | NP_666994.2 | 1 | 395 | ||
| GenBank mRNA | BC130238.1 | - | 1 | 395 | 40142117 | |
| X89681.1 | - | 1 | 17 | |||

Olfr933
Genomic Map