siRNA Details
| Gene Symbol: Olfr458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146444.1 | NP_666655.1 | 1 | 305 | ||
| GenBank mRNA | BC106796.1 | - | 1 | 305 | ||
| BC106797.1 | - | 1 | 305 | |||
| BC119415.1 | - | 1 | 305 | |||
| BC119416.1 | - | 1 | 305 | |||
| BC127219.1 | - | 1 | 300 | |||
| BC127220.1 | - | 1 | 305 | |||

Olfr458
Genomic Map