siRNA Details
| Gene Symbol: Olfr215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146446.1 | NP_666657.1 | 1 | 301 | ||
| GenBank mRNA | BC125561.1 | - | 1 | 388 | 40130076 | |
| BC125563.1 | - | 1 | 388 | 40130078 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr215| Gene Symbol: Olfr215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146446.1 | NP_666657.1 | 1 | 301 | ||
| GenBank mRNA | BC125561.1 | - | 1 | 388 | 40130076 | |
| BC125563.1 | - | 1 | 388 | 40130078 | ||