siRNA Details
| Gene Symbol: Olfr1310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146449.1 | NP_666660.1 | 1 | 765 | ||
| GenBank mRNA | BC119359.1 | - | 1 | 791 | 8734108 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1310| Gene Symbol: Olfr1310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146449.1 | NP_666660.1 | 1 | 765 | ||
| GenBank mRNA | BC119359.1 | - | 1 | 791 | 8734108 | |