siRNA Details
| Gene Symbol: Olfr164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146451.1 | NP_666662.1 | 1 | 473 | ||
| GenBank mRNA | BC145676.1 | - | 1 | 569 | 40130979 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr164| Gene Symbol: Olfr164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146451.1 | NP_666662.1 | 1 | 473 | ||
| GenBank mRNA | BC145676.1 | - | 1 | 569 | 40130979 | |