siRNA Details
| Gene Symbol: Olfr694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146452.2 | NP_666663.2 | 1 | 272 | ||
| GenBank mRNA | BC129796.1 | - | 1 | 272 | 40042951 | |
| BC129797.1 | - | 1 | 272 | 40042952 | ||
| Gene Symbol: Olfr695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146598.2 | NP_666809.1 | 3 | 525 | ||
| GenBank mRNA | BC104092.1 | - | 1 | 272 | 40042972 | |
| BC104093.1 | - | 1 | 272 | 40042973 | ||
| BC129798.1 | - | 1 | 272 | 40042986 | ||

Olfr694
Genomic Map