siRNA Details
| Gene Symbol: Olfr693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146453.2 | NP_666664.1 | 1 | 182 | ||
| GenBank mRNA | BC130248.1 | - | 1 | 185 | 40142368 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr693| Gene Symbol: Olfr693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146453.2 | NP_666664.1 | 1 | 182 | ||
| GenBank mRNA | BC130248.1 | - | 1 | 185 | 40142368 | |