siRNA Details
| Gene Symbol: Olfr1199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146458.1 | NP_666669.1 | 1 | 757 | ||
| Gene Symbol: Olfr1209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146461.2 | NP_666672.2 | 1 | 757 | ||
| GenBank mRNA | BC104269.1 | - | 1 | 757 | 40040821 | |
| BC104270.1 | - | 1 | 755 | |||

Olfr1199
Genomic Map