siRNA Details
| Gene Symbol: Olfr1202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146462.1 | NP_666673.1 | 1 | 296 | ||
| Gene Symbol: Olfr1204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146463.2 | NP_666674.2 | 1 | 296 | ||

Olfr1202
Genomic Map