siRNA Details
| Gene Symbol: Olfr1392 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146470.2 | NP_666681.1 | 1 | 98 | ||
| GenBank mRNA | BC120614.1 | - | 1 | 94 | 40129537 | |
| BC120616.1 | - | 1 | 98 | 40129539 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1392| Gene Symbol: Olfr1392 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146470.2 | NP_666681.1 | 1 | 98 | ||
| GenBank mRNA | BC120614.1 | - | 1 | 94 | 40129537 | |
| BC120616.1 | - | 1 | 98 | 40129539 | ||