siRNA Details
| Gene Symbol: Olfr1384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146472.2 | NP_666683.1 | 1 | 249 | ||
| GenBank mRNA | BC119159.1 | - | 1 | 249 | 8733908 | |
| BC120667.1 | - | 1 | 249 | 40129590 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1384| Gene Symbol: Olfr1384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146472.2 | NP_666683.1 | 1 | 249 | ||
| GenBank mRNA | BC119159.1 | - | 1 | 249 | 8733908 | |
| BC120667.1 | - | 1 | 249 | 40129590 | ||