siRNA Details
| Gene Symbol: Olfr1261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146474.1 | NP_666685.1 | 1 | 62 | ||
| GenBank mRNA | AB076075.1 | - | 1 | 62 | ||
| BC119116.1 | - | 1 | 94 | 8733865 | ||
| BC119144.1 | - | 1 | 94 | 8733893 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1261| Gene Symbol: Olfr1261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146474.1 | NP_666685.1 | 1 | 62 | ||
| GenBank mRNA | AB076075.1 | - | 1 | 62 | ||
| BC119116.1 | - | 1 | 94 | 8733865 | ||
| BC119144.1 | - | 1 | 94 | 8733893 | ||