siRNA Details
| Gene Symbol: Olfr1254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146476.1 | NP_666687.1 | 1 | 616 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1254| Gene Symbol: Olfr1254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146476.1 | NP_666687.1 | 1 | 616 | ||