siRNA Details
| Gene Symbol: Olfr183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146485.2 | NP_666696.2 | 2 | 704 | ||
| GenBank mRNA | DR065627.1 | - | 2 | 525 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr183| Gene Symbol: Olfr183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146485.2 | NP_666696.2 | 2 | 704 | ||
| GenBank mRNA | DR065627.1 | - | 2 | 525 | ||