siRNA Details
| Gene Symbol: Olfr266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146489.1 | NP_666700.1 | 1 | 646 | ||
| GenBank mRNA | BC125352.1 | - | 1 | 671 | 40129867 | |
| BC125354.1 | - | 1 | 671 | 40129869 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr266| Gene Symbol: Olfr266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146489.1 | NP_666700.1 | 1 | 646 | ||
| GenBank mRNA | BC125352.1 | - | 1 | 671 | 40129867 | |
| BC125354.1 | - | 1 | 671 | 40129869 | ||