siRNA Details
| Gene Symbol: Olfr1410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146491.1 | NP_666702.1 | 1 | 844 | ||
| GenBank mRNA | BC106991.1 | - | 1 | 844 | 40044064 | |
| BC106992.1 | - | 1 | 844 | 40044072 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1410| Gene Symbol: Olfr1410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146491.1 | NP_666702.1 | 1 | 844 | ||
| GenBank mRNA | BC106991.1 | - | 1 | 844 | 40044064 | |
| BC106992.1 | - | 1 | 844 | 40044072 | ||